Please login to your online account to display your discounted pricing

(R)-(+)-3,3'-Dibromo-1,1'-bi-2-naphthol, 98%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material (R)-(+)-3, 3'-Dibromo-1, 1'-bi-2-naphthol
CAS 111795-43-8
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C20H12Br2O2
Formula Weight 444.12
MDL Number MFCD03093629
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC384651000

  SDS
thermo fisher scientific
AC384651000
Glass bottle 100mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥97.5%
Physical Form Powder
Formula Weight 444.12
Quantity 100mg
Color White to pale yellow-beige
Specific Rotation Condition +64.00 (20.00°C c=0.2%,chloroform)
CAS Max % 100.0
Chemical Name or Material (R)-(+)-3, 3'-Dibromo-1, 1'-bi-2-naphthol
Molecular Formula C20H12Br2O2
MDL Number MFCD03093629
Infrared Spectrum Authentic
CAS Min % 97.5
Specific Rotation +60° to +68° (20°C, 589nm) (c=1, CHCl3)
Assay Percent Range 98%
CAS 111795-43-8
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Harmful if swallowed.
May cause respiratory irritation.
Causes serious eye irritation.
Packaging Glass bottle
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Safety and Handling

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
Harmful if swallowed.
May cause respiratory irritation.
Causes serious eye irritation.