Please login to your online account to display your discounted pricing

Alfa Aesar™ (R)-(-)-2-(Trifluoromethyl)pyrrolidine, 95%

Chemical Identifiers

Chemical Name or Material (R)-(-)-2-(Trifluoromethyl)pyrrolidine
CAS 119618-29-0
Molecular Formula C5H8F3N
Formula Weight 139.12
MDL Number MFCD07784180
UN Number UN2925
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2585303

 
alfa aesar™
H2585303
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH25853MD

 
alfa aesar™
H25853MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 139.12
Molecular Formula C5H8F3N
UN Number UN2925
Flash Point 26°C (79°F)
DOT Information Transport Hazard Class: 4.1; Packing Group: III; Proper Shipping Name: FLAMMABLE SOLIDS, CORROSIVE, ORGANIC, N.O.S.
Chemical Name or Material (R)-(-)-2-(Trifluoromethyl)pyrrolidine
Assay 95%
MDL Number MFCD07784180
CAS 119618-29-0
Boiling Point 110° to 112°C
Melting Point 27° to 31°C
Quantity 1g
Optical Rotation −11° (c=1 in Methanol)
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H314-H318-H228
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable solid.
Health Hazard 3 GHS P Statement
P280-P303+P361+P353-P305+P351+P338-P310
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Safety and Handling

Danger

GHS P Statement
P280-P303+P361+P353-P305+P351+P338-P310
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

GHS H Statement
H314-H318-H228
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable solid.