Learn More
(R)-2-(Trifluoromethyl)piperidine, 97%, Thermo Scientific™
Manufacturer: Thermo Fisher Scientific AC446690010
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
(R)-2-(Trifluoromethyl)piperidine | |
80.0°C (200.0 Torr) | |
C6H10F3N | |
1g | |
C1CCNC(C1)C(F)(F)F | |
153.15 | |
153.15 |
1212734-83-2 | |
96% min. (GC) | |
2r-2-trifluoromethyl piperidine, r-2-trifluoromethyl piperidine, r-2-trifluoromethylpiperidine | |
NAXDEFXCCITWEU-RXMQYKEDSA-N | |
(2R)-2-(trifluoromethyl)piperidine | |
42580053 | |
97% |
Chemical Identifiers
1212734-83-2 | |
153.15 | |
2r-2-trifluoromethyl piperidine, r-2-trifluoromethyl piperidine, r-2-trifluoromethylpiperidine | |
(2R)-2-(trifluoromethyl)piperidine |
C6H10F3N | |
NAXDEFXCCITWEU-RXMQYKEDSA-N | |
42580053 | |
C1CCNC(C1)C(F)(F)F |
Safety and Handling
GHS H Statement
Flammable liquid and vapour.
GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Wear protective gloves/protective clothing/eye p
GHS Signal Word: Danger
RUO – Research Use Only