missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
(R)-2-(Trifluoromethyl)piperidine, 97%, ACROS Organics™
Manufacturer: Thermo Fisher Scientific AC446690050
Specifications
(R)-2-(Trifluoromethyl)piperidine | |
5g | |
96% min. (GC) | |
2r-2-trifluoromethyl piperidine, r-2-trifluoromethyl piperidine, r-2-trifluoromethylpiperidine | |
C1CCNC(C1)C(F)(F)F | |
153.15 | |
153.15 |
80.0°C (200.0 Torr) | |
1212734-83-2 | |
C6H10F3N | |
NAXDEFXCCITWEU-RXMQYKEDSA-N | |
(2R)-2-(trifluoromethyl)piperidine | |
42580053 | |
97% |
Chemical Identifiers
1212734-83-2 | |
153.15 | |
2r-2-trifluoromethyl piperidine, r-2-trifluoromethyl piperidine, r-2-trifluoromethylpiperidine | |
(2R)-2-(trifluoromethyl)piperidine |
C6H10F3N | |
NAXDEFXCCITWEU-RXMQYKEDSA-N | |
42580053 | |
C1CCNC(C1)C(F)(F)F |
Safety and Handling
GHS H Statement
Flammable liquid and vapour.
GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Wear protective gloves/protective clothing/eye p
GHS Signal Word: Danger