Please login to your online account to display your discounted pricing

Alfa Aesar™ (R)-(+)-2-Phenyl-1-propanol, 98+%

$85.43 - $85.43

Chemical Identifiers

Chemical Name or Material (R)-(+)-2-Phenyl-1-propanol
CAS 19141-40-3
Molecular Formula C9H12O
Formula Weight 136.19
MDL Number MFCD00145206
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL13999MD

  SDS
alfa aesar™
L13999MD
250mg Each for $85.43
Description & Specifications

Specifications

Formula Weight 136.19
Chemical Name or Material (R)-(+)-2-Phenyl-1-propanol
Health Hazard 1 Warning
Assay ≥98%
Molecular Formula C9H12O
MDL Number MFCD00145206
CAS 19141-40-3
Boiling Point 220°C
Quantity 250mg
Flash Point 108°C (226°F)
Optical Rotation +18° (Neat)
Health Hazard 2 GHS H Statement
H302
Harmful if swallowed.
Health Hazard 3 GHS P Statement
P280f
Wear protective clothing.
Safety and Handling

Warning

GHS P Statement
P280f
Wear protective clothing.

GHS H Statement
H302
Harmful if swallowed.