missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ (R)-(-)-2-Pentanol, 97%

For synthesis of optically active products

Chemical Identifiers

CAS 31087-44-2
Molecular Formula C5H12O
Molecular Weight (g/mol) 88.15
MDL Number MFCD00065953
InChI Key JYVLIDXNZAXMDK-RXMQYKEDSA-N
Synonym r---2-pentanol, 2r-pentan-2-ol, r-2-pentanol, 2r-2-pentanol, r---2-pentanol, 2-pentanol #, r-pentan-2-ol, pubchem6721, 2-pentanol, 2r, r---2-pentanol
PubChem CID 7014876
IUPAC Name (2R)-pentan-2-ol
SMILES CCCC(C)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAJ63298MD View Documents Alfa Aesar™
J63298MD
250mg N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AAJ6329803 View Documents Alfa Aesar™
J6329803
1g N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications

Chemical Identifiers

31087-44-2
88.15
JYVLIDXNZAXMDK-RXMQYKEDSA-N
7014876
CCCC(C)O
C5H12O
MFCD00065953
r---2-pentanol, 2r-pentan-2-ol, r-2-pentanol, 2r-2-pentanol, r---2-pentanol, 2-pentanol #, r-pentan-2-ol, pubchem6721, 2-pentanol, 2r, r---2-pentanol
(2R)-pentan-2-ol

Specifications

0.81
−14° (Neat)
119°C to 120°C
250mg
C5H12O
UN1105
r---2-pentanol, 2r-pentan-2-ol, r-2-pentanol, 2r-2-pentanol, r---2-pentanol, 2-pentanol #, r-pentan-2-ol, pubchem6721, 2-pentanol, 2r, r---2-pentanol
CCCC(C)O
88.15
88.15
97%
34°C (93°F)
1.406
Colorless
31087-44-2
MFCD00065953
4652311
JYVLIDXNZAXMDK-RXMQYKEDSA-N
(2R)-pentan-2-ol
7014876
Liquid
(R)-(-)-2-Pentanol
Safety and Handling

Safety and Handling

GHS H Statement
H226-H332-H335
Flammable liquid and vapor.
Harmful if inhaled.
May cause respiratory irritation.

GHS P Statement
P210-P261-P280-P303+P361+P353-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: PENTANOLS