missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

(R)-(-)-2-Octanol, TCI America™

Manufacturer:  TCI America O014525ML

 View more versions of this product

Catalog No. O014525ML



Specifications

Specifications

(R)-(-)-2-Octanol
25mL
C8H18O
r-octan-2-ol, r---2-octanol, r-2-octanol, l-octan-2-ol, 2r-octan-2-ol, 2r-2-octanol, unii-2shb67xf6h, d-2-octanol, r---hexylmethylcarbinol, l--2-octanol
CCCCCCC(C)O
130.231
CHEBI:37871
Clear Liquid at 20°C
Colorless
5978-70-1
MFCD00064284
SJWFXCIHNDVPSH-MRVPVSSYSA-N
(2R)-octan-2-ol
80080
130.23
≥98.0% (GC)

Chemical Identifiers

5978-70-1
130.231
SJWFXCIHNDVPSH-MRVPVSSYSA-N
80080
(2R)-octan-2-ol
C8H18O
MFCD00064284
r-octan-2-ol, r---2-octanol, r-2-octanol, l-octan-2-ol, 2r-octan-2-ol, 2r-2-octanol, unii-2shb67xf6h, d-2-octanol, r---hexylmethylcarbinol, l--2-octanol
CHEBI:37871
CCCCCCC(C)O
Safety and Handling

Safety and Handling

EINECSNumber : (2)-0217

TSCA : Yes

Documents