Please login to your online account to display your discounted pricing

Alfa Aesar™ (R)-(-)-2-Nonyl isocyanate, 97%

Chemical Identifiers

Chemical Name or Material (R)-(-)-2-Nonyl isocyanate
CAS 745783-79-3
Molecular Formula C10H19NO
Formula Weight 169.27
MDL Number MFCD05664081
UN Number UN2206
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL2023603

  SDS
alfa aesar™
L2023603
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 169.27
Chemical Name or Material (R)-(-)-2-Nonyl isocyanate
Health Hazard 3 GHS P Statement
P284a-P261-P280-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
CAS 745783-79-3
Health Hazard 1 Danger
MDL Number MFCD05664081
Molecular Formula C10H19NO
Recommended Storage 2° to 8°C
Assay 97%
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: ISOCYANATES, TOXIC, N.O.S.
Quantity 1g
UN Number UN2206
Health Hazard 2 GHS H Statement
H331-H334-H302-H315-H319-H335
Toxic if inhaled.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Safety and Handling

Danger

GHS P Statement
P284a-P261-P280-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H331-H334-H302-H315-H319-H335
Toxic if inhaled.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

2° to 8°C