missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

(R)-2-Methylpyrrolidine Hydrochloride, TCI America™

$122.22 - $366.67

Chemical Identifiers

CAS 135324-85-5
Molecular Formula C5H12ClN
Molecular Weight (g/mol) 121.608
MDL Number MFCD06796610
InChI Key JNEIFWYJFOEKIM-NUBCRITNSA-N
PubChem CID 11147704
IUPAC Name (2R)-2-methylpyrrolidine;hydrochloride
SMILES CC1CCCN1.Cl
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
M27471G View Documents TCI America
M27471G
1g Each for $366.67
 
M2747200MG View Documents TCI America
M2747200MG
200mg Each for $122.22
 
Specifications

Chemical Identifiers

135324-85-5
121.608
JNEIFWYJFOEKIM-NUBCRITNSA-N
(2R)-2-methylpyrrolidine;hydrochloride
C5H12ClN
MFCD06796610
11147704
CC1CCCN1.Cl

Specifications

White-Yellow
1g
C5H12ClN
JNEIFWYJFOEKIM-NUBCRITNSA-N
(2R)-2-methylpyrrolidine;hydrochloride
11147704
Crystal-Powder at 20°C
(R)-2-Methylpyrrolidine Hydrochloride
75°C
135324-85-5
MFCD06796610
CC1CCCN1.Cl
121.608
121.61
≥97.0% (N,T)
Safety and Handling

Safety and Handling

TSCA : No