missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
(R)-(-)-2-Methylpiperazine, 98%, ACROS Organics™
Supplier: Thermo Scientific Chemicals 356270050
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
(R)-(-)-2-Methylpiperazine | |
155.0°C to 156.0°C | |
Authentic | |
Glass bottle | |
5g | |
− 18.00 | |
Solubility in water: soluble. | |
CC1CNCCN1 | |
100.16 | |
100.16 |
75336-86-6 | |
65°C | |
95% min. (GC) | |
C5H12N2 | |
− 18.00 (20.00°C c=1,toluene) | |
r---2-methylpiperazine, r-2-methylpiperazine, 2r-2-methylpiperazine, r-mp, piperazine, 2-methyl-, 2r, r-2-methyl-piperazine, 2-methylpiperazine, r, unii-p4xu643r8w | |
JOMNTHCQHJPVAZ-RXMQYKEDSA-N | |
(2R)-2-methylpiperazine | |
7330434 | |
95% |
Chemical Identifiers
75336-86-6 | |
100.16 | |
r---2-methylpiperazine, r-2-methylpiperazine, 2r-2-methylpiperazine, r-mp, piperazine, 2-methyl-, 2r, r-2-methyl-piperazine, 2-methylpiperazine, r, unii-p4xu643r8w | |
(2R)-2-methylpiperazine |
C5H12N2 | |
JOMNTHCQHJPVAZ-RXMQYKEDSA-N | |
7330434 | |
CC1CNCCN1 |
Safety and Handling
GHS Signal Word: Danger
RUO â Research Use Only