Please login to your online account to display your discounted pricing

(R)-(-)-2-Methylpiperazine, ACROS Organics™


Quantity 1g

Manufacturer: thermo fisher scientific  356270010

 View more versions of this product

Catalog No. AC356270010

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥97.5% (GC)
Physical Form Crystalline Powder, Crystals and/or Chunks
Formula Weight 100.16
Quantity 1g
Color White to yellow
Melting Point 86° to 91°C
Specific Rotation Condition − 18.00 (20.00°C c=1,toluene)
Specific Rotation −15° to −21° (20°C, 589nm) (c=1, toluene)
Assay Percent Range 98%
CAS 75336-86-6
Chemical Name or Material (R)-(-)-2-Methylpiperazine
Infrared Spectrum Authentic
Molecular Formula C5H12N2

Specifications

Chemical Name or Material (R)-(-)-2-Methylpiperazine
Assay ≥97.5% (GC)
Assay Percent Range 98%
Molecular Formula C5H12N2
Formula Weight 100.16