Learn More
(R)-(+)-2-Methyl-CBS-oxazaborolidine, 1M solution in toluene, Thermo Scientific™
Chemical Identifiers
CAS | 112022-83-0 |
---|---|
Molecular Formula | C18H20BNO |
Molecular Weight (g/mol) | 277.17 |
InChI Key | VMKAFJQFKBASMU-QGZVFWFLSA-N |
Synonym | r-2-methyl-cbs-oxazaborolidine, r-methyl oxazaborolidine, r-+-2-methyl-cbs-oxazaborolidine, r-5,5-diphenyl-2-methyl-3,4-propano-1,3,2-oxazaborolidine, r-1-methyl-3,3-diphenylhexahydropyrrolo 1,2-c 1,3,2 oxazaborole, r-me-cbs catalyst, r-3,3-diphenyl-1-methylpyrrolidino 1,2-c-1,3,2-oxazaborole, r-me cbs, r-tetrahydro-1-methyl-3,3-diphenyl-1h,3h-pyrrolo 1,2-c 1,3,2 oxazaborole, corey's catalyst |
PubChem CID | 9838490 |
IUPAC Name | (3aR)-1-methyl-3,3-diphenyl-3a,4,5,6-tetrahydropyrrolo[1,2-c][1,3,2]oxazaborole |
SMILES | B1(N2CCCC2C(O1)(C3=CC=CC=C3)C4=CC=CC=C4)C |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC369780050
|
Thermo Scientific Chemicals
369780050 |
5mL | Glass bottle |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
AC369780250
|
Thermo Scientific Chemicals
369780250 |
25mL | Glass bottle |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
112022-83-0 | |
277.17 | |
r-2-methyl-cbs-oxazaborolidine, r-methyl oxazaborolidine, r-+-2-methyl-cbs-oxazaborolidine, r-5,5-diphenyl-2-methyl-3,4-propano-1,3,2-oxazaborolidine, r-1-methyl-3,3-diphenylhexahydropyrrolo 1,2-c 1,3,2 oxazaborole, r-me-cbs catalyst, r-3,3-diphenyl-1-methylpyrrolidino 1,2-c-1,3,2-oxazaborole, r-me cbs, r-tetrahydro-1-methyl-3,3-diphenyl-1h,3h-pyrrolo 1,2-c 1,3,2 oxazaborole, corey's catalyst | |
(3aR)-1-methyl-3,3-diphenyl-3a,4,5,6-tetrahydropyrrolo[1,2-c][1,3,2]oxazaborole |
C18H20BNO | |
VMKAFJQFKBASMU-QGZVFWFLSA-N | |
9838490 | |
B1(N2CCCC2C(O1)(C3=CC=CC=C3)C4=CC=CC=C4)C |
Specifications
112022-83-0,108-88-3 | |
111.0°C | |
Glass bottle | |
5mL | |
r-2-methyl-cbs-oxazaborolidine, r-methyl oxazaborolidine, r-+-2-methyl-cbs-oxazaborolidine, r-5,5-diphenyl-2-methyl-3,4-propano-1,3,2-oxazaborolidine, r-1-methyl-3,3-diphenylhexahydropyrrolo 1,2-c 1,3,2 oxazaborole, r-me-cbs catalyst, r-3,3-diphenyl-1-methylpyrrolidino 1,2-c-1,3,2-oxazaborole, r-me cbs, r-tetrahydro-1-methyl-3,3-diphenyl-1h,3h-pyrrolo 1,2-c 1,3,2 oxazaborole, corey's catalyst | |
VMKAFJQFKBASMU-QGZVFWFLSA-N | |
(3aR)-1-methyl-3,3-diphenyl-3a,4,5,6-tetrahydropyrrolo[1,2-c][1,3,2]oxazaborole | |
9838490 | |
(R)-(+)-2-Methyl-CBS-oxazaborolidine |
0.9500g/mL | |
4°C | |
C18H20BNO | |
0.95 | |
Solubility in water: reacts | |
B1(N2CCCC2C(O1)(C3=CC=CC=C3)C4=CC=CC=C4)C | |
277.17 | |
277.17 |
Safety and Handling
GHS H Statement
Causes serious eye damage.
Harmful if swallowed.
May be fatal if swallowed and enters airways.
May cause drowsiness or dizziness.
May cause damage to organs through prolonged or repeated exposure.
Caus
GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF INHALED: Remov
GHS Signal Word: Danger
RUO â Research Use Only