Learn More
(R)-(+)-2-Methyl-2-propanesulfinamide, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals H2772406
Description
(R)-(+)-2-Methyl-2-propanesulfinamide is a chiral ligand, which is used in pharmaceutical compositions. Further, it is used in the preparation of beta-chloro sulfinamides in the synthesis of chiral azridines. It is involved in the preparation of organocatalyst for enantioselective reduction of imines. It serves as a reagent for synthesizing chiral amines. In addition to this, it is converted into P,N-sulfinyl imine ligands through condensation with aldehydes and ketones which undergoes iridium-catalyzed asymmetric hydrogenation of olefins.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications(R)-(+)-2-Methyl-2-propanesulfinamide is a chiral ligand, which is used in pharmaceutical compositions. Further, it is used in the preparation of beta-chloro sulfinamides in the synthesis of chiral azridines. It is involved in the preparation of organocatalyst for enantioselective reduction of imines. It serves as a reagent for synthesizing chiral amines. In addition to this, it is converted into P,N-sulfinyl imine ligands through condensation with aldehydes and ketones which undergoes iridium-catalyzed asymmetric hydrogenation of olefins.
Solubility
Soluble in chloroform, methanol, tetrahydrofuran, dichloromethane, dimethyl sulfoxide and most organic solvents.
Notes
Incompatible with strong oxidizing agents, strong acids and bases. Store in a cool place.
Specifications
(R)-(+)-2-Methyl-2-propanesulfinamide | |
∼100°C to 105°C | |
MFCD05861479 | |
r-+-2-methyl-2-propanesulfinamide, r-2-methylpropane-2-sulfinamide, r-+-t-butylsulfinamide, r-+-tert-butanesulfinamide, r-+-tert-butylsulfinamide, r-2-methyl-2-propanesulfinamide, r-tert-butanesulfinamide, r-+-tert-butyl sulphinamide, r-tert-butylsulfinamide, r-+-tert-butyl sulfinamide | |
CESUXLKAADQNTB-UHFFFAOYNA-N | |
2-methylpropane-2-sulfinamide | |
10964479 | |
98% |
196929-78-9 | |
C4H11NOS | |
5 g | |
Soluble in chloroform,methanol,tetrahydrofuran,dichloromethane,dimethyl sulfoxide and most organic solvents. | |
CC(C)(C)S(N)=O | |
121.20 | |
121.2 |
Chemical Identifiers
196929-78-9 | |
121.20 | |
CESUXLKAADQNTB-UHFFFAOYNA-N | |
10964479 | |
CC(C)(C)S(N)=O |
C4H11NOS | |
MFCD05861479 | |
r-+-2-methyl-2-propanesulfinamide, r-2-methylpropane-2-sulfinamide, r-+-t-butylsulfinamide, r-+-tert-butanesulfinamide, r-+-tert-butylsulfinamide, r-2-methyl-2-propanesulfinamide, r-tert-butanesulfinamide, r-+-tert-butyl sulphinamide, r-tert-butylsulfinamide, r-+-tert-butyl sulfinamide | |
2-methylpropane-2-sulfinamide |
Safety and Handling
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
Recommended Storage : Keep cold
RUO – Research Use Only