Please login to your online account to display your discounted pricing

Alfa Aesar™ (R)-(-)-2-Methyl-2,4-pentanediol, 98+%

$47.26 - $116.36

Chemical Identifiers

Chemical Name or Material (R)-(-)-2-Methyl-2,4-pentanediol
CAS 99210-90-9
Molecular Formula C6H14O2
Formula Weight 118.18
MDL Number MFCD00064275
Beilstein 6051650
Synonym (R)-(-)-Hexylene glycol
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1452303

 
alfa aesar™
L1452303
1g Each for $116.36

AAL14523MD

 
alfa aesar™
L14523MD
250mg Each for $47.26
Description & Specifications

Specifications

Formula Weight 118.18
Chemical Name or Material (R)-(-)-2-Methyl-2,4-pentanediol
Molecular Formula C6H14O2
MDL Number MFCD00064275
CAS 99210-90-9
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H227
Causes skin irritation.
Causes serious eye irritation.
Combustible liquid.
Health Hazard 3 GHS P Statement
P210-P280-P305+P351+P338-P362-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Take off contaminated clothing and wash before reuse.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Assay ≥98%
Beilstein 6051650
Boiling Point 197°C
Synonym (R)-(-)-Hexylene glycol
Quantity 1g
Flash Point 93°C (199°F)
Optical Rotation −21° (Neat)

Applications
(R)-(-)-2-Methyl-2,4-pentanediol is used as a chiral reagent. Also used as intermediate for pharmaceutical.

Solubility
Fully miscible with water.

Notes
Hygroscopic, store away from water/moisture. Store in cool. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.

Safety and Handling

Warning

GHS P Statement
P210-P280-P305+P351+P338-P362-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Take off contaminated clothing and wash before reuse.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H227
Causes skin irritation.
Causes serious eye irritation.
Combustible liquid.