Please login to your online account to display your discounted pricing

Alfa Aesar™ (R)-(-)-2-Methoxy-2-phenylethanol, 98+%

$74.28 - $183.82

Chemical Identifiers

Chemical Name or Material (R)-(-)-2-Methoxy-2-phenylethanol
CAS 17628-72-7
Molecular Formula C9H12O2
Formula Weight 152.19
MDL Number MFCD00011550
Beilstein 2437247
Synonym (R)-(-)-beta-Methoxyphenethyl alcohol
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1404403

  SDS
alfa aesar™
L1404403
1g Each for $183.82

AAL14044MD

  SDS
alfa aesar™
L14044MD
250mg Each for $74.28
Description & Specifications

Specifications

Formula Weight 152.19
Chemical Name or Material (R)-(-)-2-Methoxy-2-phenylethanol
Molecular Formula C9H12O2
CAS 17628-72-7
MDL Number MFCD00011550
Assay ≥98%
Boiling Point 65°C (0.1mmHg)
Beilstein 2437247
Synonym (R)-(-)-beta-Methoxyphenethyl alcohol
Quantity 1g
Optical Rotation −133° (c=1 in Acetone)
Flash Point 97°C (206°F)