missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

(R)-2-Isopropylsuccinic acid-1-methyl ester, 95%, (98% ee), ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC363060050

 View more versions of this product

Catalog No. AC363060050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Specifications

(R)-2-Isopropylsuccinic acid-1-methyl ester
Authentic
Yellow
98% ee
C8H14O4
2r-2-isopropylsuccinic acid 1-methyl ester, 3r-3-methoxycarbonyl-4-methylpentanoic acid, butanedioic acid,2-1-methylethyl-, 1-methyl ester, 2r, r---2-isopropylsuccinic acid 1-methyl ester, r-2-isopropyl-succinic acid-1-methyl ester, r-2-isopropylsuccinic acid-1-methyl ester, r-3-methoxycarbonyl-4-methylpentanoic acid
CC(C)C(CC(=O)O)C(=O)OC
174.19
174.19
95%,(98% ee)
97.5% min. (GC)
Glass Bottle
5g
220498-08-8
MFCD01091030
VHOGSWBAPDPWTG-ZCFIWIBFSA-N
(3R)-3-methoxycarbonyl-4-methylpentanoic acid
5702634
Viscous Liquid

Chemical Identifiers

220498-08-8
174.19
VHOGSWBAPDPWTG-ZCFIWIBFSA-N
5702634
CC(C)C(CC(=O)O)C(=O)OC
C8H14O4
MFCD01091030
2r-2-isopropylsuccinic acid 1-methyl ester, 3r-3-methoxycarbonyl-4-methylpentanoic acid, butanedioic acid,2-1-methylethyl-, 1-methyl ester, 2r, r---2-isopropylsuccinic acid 1-methyl ester, r-2-isopropyl-succinic acid-1-methyl ester, r-2-isopropylsuccinic acid-1-methyl ester, r-3-methoxycarbonyl-4-methylpentanoic acid
(3R)-3-methoxycarbonyl-4-methylpentanoic acid
SDS Documents