missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

(R)-2-Isobutylsuccinic acid-1-methyl ester, 95% (98% e.e.), ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 363055000

 View more versions of this product

Catalog No. AC363055000

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

(R)-2-Isobutylsuccinic acid-1-methyl ester
95%, (98% e.e.)
97.5% min. (GC)
≥94%
188.22
(R)-3-Methoxycarbonyl-5-methylhexanoic acid
Glass Ampule with screw-cap
Yellow
500mg
130165-76-3
Viscous Liquid
C9H16O4
MFCD01091032
Authentic
+18° to +21° (25°C, 589nm) (c=1, CH3OH)

Chemical Identifiers

130165-76-3
188.22
(R)-3-Methoxycarbonyl-5-methylhexanoic acid
≥94%
C9H16O4
MFCD01091032
(R)-2-Isobutylsuccinic acid-1-methyl ester, 95%, (98% e.e.)

Safety and Handling

SDS & Safety Documents

GHS H Statement:
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

TSCA : TSCA