Learn More
(R)-2-Isobutylsuccinic acid-1-methyl ester, 95%, (98% ee), Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 363050010
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
(R)-2-Isobutylsuccinic acid-1-methyl ester | |
130165-76-3 | |
97.5% min. (GC) | |
Glass bottle | |
MFCD01091032 | |
+18° to +21° (25°C, 589nm) (c=1, CH3OH) | |
AJNNFMRCMKGYHA-SSDOTTSWSA-N | |
(3R)-3-methoxycarbonyl-5-methylhexanoic acid | |
5702633 | |
95%,(98% ee) |
95%, (98% e.e.) | |
Yellow | |
Authentic | |
C9H16O4 | |
1 g | |
r-2-isobutylsuccinic acid-1-methyl ester, 3r-3-methoxycarbonyl-5-methylhexanoic acid, r-3-methoxycarbonyl-5-methylhexanoic acid, butanedioic acid,2-2-methylpropyl-, 1-methyl ester, 2r, r-2-isobutyl succinic acid 1-methyl ester, r-+-2-isobutylsuccinic acid 1-methyl ester | |
CC(C)CC(CC(=O)O)C(=O)OC | |
188.22 | |
188.22 | |
Viscous Liquid |
Chemical Identifiers
130165-76-3 | |
188.22 | |
AJNNFMRCMKGYHA-SSDOTTSWSA-N | |
5702633 | |
CC(C)CC(CC(=O)O)C(=O)OC |
C9H16O4 | |
MFCD01091032 | |
r-2-isobutylsuccinic acid-1-methyl ester, 3r-3-methoxycarbonyl-5-methylhexanoic acid, r-3-methoxycarbonyl-5-methylhexanoic acid, butanedioic acid,2-2-methylpropyl-, 1-methyl ester, 2r, r-2-isobutyl succinic acid 1-methyl ester, r-+-2-isobutylsuccinic acid 1-methyl ester | |
(3R)-3-methoxycarbonyl-5-methylhexanoic acid |
Safety and Handling
GHS H Statement
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove
GHS Signal Word: Warning
TSCA : TSCA
RUO – Research Use Only