missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
(R)-2-Hydroxymethylmorpholine hydrochloride, 95%, Thermo Scientific Chemicals
$826.64 - $826.64
Chemical Identifiers
CAS | 1436436-17-7 |
---|---|
Molecular Formula | C5H12ClNO2 |
MDL Number | MFCD11052541 |
Synonym | r-morpholin-2-ylmethanol hydrochloride, r-2-hydroxymethylmorpholine hcl, r-morpholin-2-yl methanol hydrochloride, r-2-hydroxymethylmorpholine hydrochloride, r-morpholin-2-ylmethanol-hcl, r-2-morpholinemethanol hcl, r-2-morpholinemethanol hydrochloride, 2r-morpholin-2-ylmethanol hydrochloride, r-2-hydroxymethyl-morpholine hydrochloride, r-2-hydroxymethyl morpholine hydrochloride |
PubChem CID | 42614735 |
IUPAC Name | [(2R)-morpholin-2-yl]methanol;hydrochloride |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAH6400303
|
Thermo Scientific Chemicals
H6400303 |
1 g |
Each for $826.64
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
1436436-17-7 | |
MFCD11052541 | |
42614735 |
C5H12ClNO2 | |
r-morpholin-2-ylmethanol hydrochloride, r-2-hydroxymethylmorpholine hcl, r-morpholin-2-yl methanol hydrochloride, r-2-hydroxymethylmorpholine hydrochloride, r-morpholin-2-ylmethanol-hcl, r-2-morpholinemethanol hcl, r-2-morpholinemethanol hydrochloride, 2r-morpholin-2-ylmethanol hydrochloride, r-2-hydroxymethyl-morpholine hydrochloride, r-2-hydroxymethyl morpholine hydrochloride | |
[(2R)-morpholin-2-yl]methanol;hydrochloride |
Specifications
1436436-17-7 | |
MFCD11052541 | |
r-morpholin-2-ylmethanol hydrochloride, r-2-hydroxymethylmorpholine hcl, r-morpholin-2-yl methanol hydrochloride, r-2-hydroxymethylmorpholine hydrochloride, r-morpholin-2-ylmethanol-hcl, r-2-morpholinemethanol hcl, r-2-morpholinemethanol hydrochloride, 2r-morpholin-2-ylmethanol hydrochloride, r-2-hydroxymethyl-morpholine hydrochloride, r-2-hydroxymethyl morpholine hydrochloride | |
C1COC(CN1)CO.Cl | |
153.606 | |
153.61 | |
Solid |
C5H12ClNO2 | |
1 g | |
NBGXGDBTUJNTKJ-NUBCRITNSA-N | |
[(2R)-morpholin-2-yl]methanol;hydrochloride | |
42614735 | |
95% | |
(R)-2-Hydroxymethylmorpholine hydrochloride |
Safety and Handling
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only