missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

(R)-2-Hydroxy-4-phenylbutyric Acid 98.0+%, TCI America™

$77.36 - $266.63

Chemical Identifiers

CAS 29678-81-7
Molecular Formula C10H12O3
Molecular Weight (g/mol) 180.203
MDL Number MFCD00192219
InChI Key JNJCEALGCZSIGB-SECBINFHSA-N
Synonym 2 r-hydroxy-4-phenylbutanoic acid, 2r-2-hydroxy-4-phenylbutanoic acid, 2r-hydroxy-4-phenylbutanoic acid, pubchem5687, r---2-hydroxy-4-phenylbutyric acid, r-2-hydroxy-4-phenyl-butyric acid, r-2-hydroxy-4-phenylbutanoic acid, r-2-hydroxy-4-phenylbutyric acid, r-2-hydroxy-4-phenylbutyricacid
PubChem CID 2759328
IUPAC Name (2R)-2-hydroxy-4-phenylbutanoic acid
SMILES C1=CC=C(C=C1)CCC(C(=O)O)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
H13381G View Documents Tci America
H13381G
1g Each for $77.36
 
H13385G View Documents Tci America
H13385G
5g Each for $266.63
 
Specifications

Chemical Identifiers

29678-81-7
180.203
JNJCEALGCZSIGB-SECBINFHSA-N
2759328
C1=CC=C(C=C1)CCC(C(=O)O)O
C10H12O3
MFCD00192219
2 r-hydroxy-4-phenylbutanoic acid, 2r-2-hydroxy-4-phenylbutanoic acid, 2r-hydroxy-4-phenylbutanoic acid, pubchem5687, r---2-hydroxy-4-phenylbutyric acid, r-2-hydroxy-4-phenyl-butyric acid, r-2-hydroxy-4-phenylbutanoic acid, r-2-hydroxy-4-phenylbutyric acid, r-2-hydroxy-4-phenylbutyricacid
(2R)-2-hydroxy-4-phenylbutanoic acid

Specifications

White
1g
C10H12O3
2 r-hydroxy-4-phenylbutanoic acid, 2r-2-hydroxy-4-phenylbutanoic acid, 2r-hydroxy-4-phenylbutanoic acid, pubchem5687, r---2-hydroxy-4-phenylbutyric acid, r-2-hydroxy-4-phenyl-butyric acid, r-2-hydroxy-4-phenylbutanoic acid, r-2-hydroxy-4-phenylbutyric acid, r-2-hydroxy-4-phenylbutyricacid
C1=CC=C(C=C1)CCC(C(=O)O)O
180.203
180.20
≥98.0% (T)
117°C
29678-81-7
MFCD00192219
JNJCEALGCZSIGB-SECBINFHSA-N
(2R)-2-hydroxy-4-phenylbutanoic acid
2759328
Crystal-Powder at 20°C
(R)-2-Hydroxy-4-phenylbutyric Acid
Safety and Handling

Safety and Handling

EINECSNumber : (3)-4133

TSCA : No