missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ (R)-(-)-2-Hexyl isocyanate, 95%

Chemical Identifiers

CAS 745783-77-1
Molecular Formula C7H13NO
Formula Weight 127.19
MDL Number MFCD05664079
UN Number UN3080
Beilstein 8253639
Chemical Name or Material (R)-(-)-2-Hexyl isocyanate
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Qty  
Catalog Number Mfr. No. Quantity Price Qty  
AAL2023403 SDS Alfa Aesar™
L2023403
1g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

745783-77-1
127.19
UN3080
(R)-(-)-2-Hexyl isocyanate
C7H13NO
MFCD05664079
8253639

Specifications

745783-77-1
127.19
UN3080
1g
(R)-(-)-2-Hexyl isocyanate
C7H13NO
MFCD05664079
8253639
47°C (117°F)
95%

Safety and Handling

GHS H Statement
H330-H334-H226-H302-H315-H319-H335
Fatal if inhaled.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Flammable liquid and vapor.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P210-P303+P361+P353-P304+P340-P305+P351+P338-P320-P330-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Specific treatment is urgent (see label).
Rinse mouth.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: ISOCYANATES, TOXIC, FLAMMABLE, N.O.S.

Recommended Storage : 2° to 8°C