missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ (R)-2-(Fmoc-amino)-4-aminobutyric acid, 95%

$141.42 - $422.54

Chemical Identifiers

CAS 201484-12-0
Molecular Formula C19H20N2O4
Molecular Weight (g/mol) 340.379
MDL Number MFCD00237018
InChI Key ZZDRDGKSMGGBDI-QGZVFWFLSA-N
Synonym fmoc-d-dab-oh, r-2-9h-fluoren-9-yl methoxy carbonyl amino-4-aminobutanoic acid, 2r-4-amino-2-9h-fluoren-9-ylmethoxy carbonyl amino butanoic acid, 2r-4-amino-2-9h-fluoren-9-ylmethoxycarbonylamino butanoic acid, n-a-fmoc-d-2,4-diaminobutyric acid, 2r-4-amino-2-fmoc-amino butanoic acid, n-alpha-9-fluorenylmethyloxycarbonyl-d-2,4-diaminobutyric acid, r-2-9h-fluoren-9-yl methoxy carbonyl amino-4-aminobutanoicacid
PubChem CID 7019710
IUPAC Name (2R)-4-amino-2-(9H-fluoren-9-ylmethoxycarbonylamino)butanoic acid
SMILES C1=CC=C2C(=C1)C(C3=CC=CC=C32)COC(=O)NC(CCN)C(=O)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH66094MD View Documents Alfa Aesar™
H66094MD
250mg Each for $141.42
 
AAH6609403 View Documents Alfa Aesar™
H6609403
1g Each for $422.54
 
Specifications

Chemical Identifiers

201484-12-0
340.379
ZZDRDGKSMGGBDI-QGZVFWFLSA-N
7019710
C1=CC=C2C(=C1)C(C3=CC=CC=C32)COC(=O)NC(CCN)C(=O)O
C19H20N2O4
MFCD00237018
fmoc-d-dab-oh, r-2-9h-fluoren-9-yl methoxy carbonyl amino-4-aminobutanoic acid, 2r-4-amino-2-9h-fluoren-9-ylmethoxy carbonyl amino butanoic acid, 2r-4-amino-2-9h-fluoren-9-ylmethoxycarbonylamino butanoic acid, n-a-fmoc-d-2,4-diaminobutyric acid, 2r-4-amino-2-fmoc-amino butanoic acid, n-alpha-9-fluorenylmethyloxycarbonyl-d-2,4-diaminobutyric acid, r-2-9h-fluoren-9-yl methoxy carbonyl amino-4-aminobutanoicacid
(2R)-4-amino-2-(9H-fluoren-9-ylmethoxycarbonylamino)butanoic acid

Specifications

White
201484-12-0
MFCD00237018
ZZDRDGKSMGGBDI-QGZVFWFLSA-N
(2R)-4-amino-2-(9H-fluoren-9-ylmethoxycarbonylamino)butanoic acid
7019710
Crystalline Powder
(R)-2-(Fmoc-amino)-4-aminobutyric acid
250mg
C19H20N2O4
fmoc-d-dab-oh, r-2-9h-fluoren-9-yl methoxy carbonyl amino-4-aminobutanoic acid, 2r-4-amino-2-9h-fluoren-9-ylmethoxy carbonyl amino butanoic acid, 2r-4-amino-2-9h-fluoren-9-ylmethoxycarbonylamino butanoic acid, n-a-fmoc-d-2,4-diaminobutyric acid, 2r-4-amino-2-fmoc-amino butanoic acid, n-alpha-9-fluorenylmethyloxycarbonyl-d-2,4-diaminobutyric acid, r-2-9h-fluoren-9-yl methoxy carbonyl amino-4-aminobutanoicacid
C1=CC=C2C(=C1)C(C3=CC=CC=C32)COC(=O)NC(CCN)C(=O)O
340.379
340.38
95%
Safety and Handling

Safety and Handling

storageNote1 : -30° to -10°C