Please login to your online account to display your discounted pricing

Alfa Aesar™ (R)-(+)-2'-Diphenylphosphino-1,1'-binaphth-2-ol, 96%

$190.86 - $581.85

Chemical Identifiers

Chemical Name or Material (R)-(+)-2'-Diphenylphosphino-1,1'-binaphth-2-ol
CAS 149917-88-4
Molecular Formula C32H23OP
Formula Weight 454.51
MDL Number MFCD01166485
Synonym (R)-(+)-2-Diphenylphosphino-2'-hydroxy-1,1'-binaphthyl
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH27032MD

 
alfa aesar™
H27032MD
250mg Each for $581.85

AAH27032MF

 
alfa aesar™
H27032MF
50mg Each for $190.86
Description & Specifications

Specifications

Formula Weight 454.51
Chemical Name or Material (R)-(+)-2'-Diphenylphosphino-1,1'-binaphth-2-ol
Assay 96%
Molecular Formula C32H23OP
MDL Number MFCD01166485
CAS 149917-88-4
Synonym (R)-(+)-2-Diphenylphosphino-2'-hydroxy-1,1'-binaphthyl
Health Hazard 1 Warning
Quantity 250mg
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.