Please login to your online account to display your discounted pricing

(R)-2-(Cyclohexylmethyl)succinic acid-1-methyl ester, 95%, 98% e.e., ACROS Organics™

$1,090.85 - $1,090.85

Chemical Identifiers

Chemical Name or Material (R)-2-(Cyclohexylmethyl)succinic acid-1-methyl ester
Name Note 95%, (98% e.e.)
CAS 130165-88-7
CAS Min % 94.0
CAS Max % 100.0
Assay ≥94%
Assay Percent Range 95%
Molecular Formula C12H20O4
Formula Weight 228.28
MDL Number MFCD01311785
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC363030010

  SDS
thermo fisher scientific
363030010
Glass bottle 1g Each for $1,090.85
Description & Specifications

Specifications

Assay ≥94%
Physical Form Viscous Liquid
Formula Weight 228.28
Quantity 1g
Color Yellow
TSCA TSCA
CAS Min % 94.0
CAS Max % 100.0
CAS 130165-88-7
Synonym (R)-4-Cyclohexyl-3-(methoxycarbonyl)butyric acid
Enantiomeric Excess 97.5% min. (GC)
Chemical Name or Material (R)-2-(Cyclohexylmethyl)succinic acid-1-methyl ester
Infrared Spectrum Authentic
Packaging Glass bottle
Name Note 95%, (98% e.e.)
Assay Percent Range 95%
Molecular Formula C12H20O4
MDL Number MFCD01311785