Please login to your online account to display your discounted pricing

(R)-2-Cyclohexyl succinic acid-1-methyl ester, 95%, 98% e.e., ACROS Organics™


Quantity 500mg
Packaging Glass bottle

Manufacturer: thermo fisher scientific  363045000

 View more versions of this product

Catalog No. AC363045000

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥94%
Physical Form Needle-Like Crystals
Formula Weight 214.26
Quantity 500mg
Color Pale yellow
TSCA TSCA
CAS Min % 94.0
CAS Max % 100.0
Chemical Name or Material (R)-2-Cyclohexyl succinic acid-1-methyl ester
Infrared Spectrum Authentic
Name Note 95%, (98% e.e.)
Assay Percent Range 95%
Molecular Formula C11 H18 O4
MDL Number MFCD01091028
CAS 220498-07-7
Packaging Glass bottle
Synonym (R)-3-Cyclohexyl-3-(methoxycarbonyl)propionic acid
Enantiomeric Excess 97.5% min. (GC)

Specifications

Chemical Name or Material (R)-2-Cyclohexyl succinic acid-1-methyl ester
Name Note 95%, (98% e.e.)
CAS 220498-07-7
CAS Min % 94.0
CAS Max % 100.0
Assay ≥94%
Assay Percent Range 95%
Molecular Formula C11 H18 O4
Formula Weight 214.26
MDL Number MFCD01091028
Synonym (R)-3-Cyclohexyl-3-(methoxycarbonyl)propionic acid