Please login to your online account to display your discounted pricing

(R)-2-Cyclohexyl succinic acid-1-methyl ester, 95%, 98% e.e., ACROS Organics™

Chemical Identifiers

Chemical Name or Material (R)-2-Cyclohexyl succinic acid-1-methyl ester
Name Note 95%, (98% e.e.)
CAS 220498-07-7
CAS Min % 94.0
CAS Max % 100.0
Assay ≥94%
Assay Percent Range 95%
Molecular Formula C11 H18 O4
Formula Weight 214.26
MDL Number MFCD01091028
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC363040010

  SDS
thermo fisher scientific
363040010
Glass bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC363045000

  SDS
thermo fisher scientific
363045000
Glass bottle 500mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥94%
Physical Form Needle-Like Crystals
Formula Weight 214.26
Quantity 1g
Color Pale yellow
TSCA TSCA
CAS Min % 94.0
CAS Max % 100.0
Chemical Name or Material (R)-2-Cyclohexyl succinic acid-1-methyl ester
Molecular Formula C11 H18 O4
MDL Number MFCD01091028
CAS 220498-07-7
Synonym (R)-3-Cyclohexyl-3-(methoxycarbonyl)propionic acid
Enantiomeric Excess 97.5% min. (GC)
Infrared Spectrum Authentic
Packaging Glass bottle
Name Note 95%, (98% e.e.)
Assay Percent Range 95%