missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

(R)-(-)-2-Chloromandelic acid, 99%, ACROS Organics™

Chemical Identifiers

CAS 52950-18-2
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 99%
Molecular Formula C8 H7 Cl O3
Formula Weight 186.59
MDL Number MFCD00798429
Chemical Name or Material (R)-(-)-2-Chloromandelic acid, 99%
Assay ≥98.5%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
AC333210010 SDS Thermo Fisher Scientific
333210010
Glass bottle 1g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

52950-18-2
100.0
C8 H7 Cl O3
MFCD00798429
≥98.5%
98.5
99%
186.59
(R)-(-)-2-Chloromandelic acid, 99%

Specifications

Flakes or Crystalline Powder
1g
118° to 121°C
-123° to -127° (20°C, 589nm) (c=3, water)
MFCD00798429
C8 H7 Cl O3
Glass bottle
(R)-(-)-2-Chloromandelic acid, 99%
186.59
White to off-white
100.0
99%
98.5
52950-18-2
Authentic
≥98.5%

Safety and Handling

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

TSCA : TSCA