Please login to your online account to display your discounted pricing

(R)-2-Butylsuccinic acid-1-methyl ester, 95%, 98% e.e., ACROS Organics™


Quantity 500mg
Packaging Glass Ampule with Screw-cap

Manufacturer: thermo fisher scientific  363085000

 View more versions of this product

Catalog No. AC363085000

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥94%
Physical Form Liquid
Formula Weight 188.2
Quantity 500mg
Color Clear orange
Packaging Glass Ampule with Screw-cap
TSCA TSCA
CAS Min % 94.0
CAS Max % 100.0
Chemical Name or Material (R)-2-Butylsuccinic acid-1-methyl ester
Name Note 95%, (98% e.e.)
Molecular Formula C9 H16 O4
Infrared Spectrum Authentic
Enantiomeric Excess 97.5% min. (GC)
Assay Percent Range 95%
MDL Number MFCD03427672
CAS 850411-21-1

Specifications

Chemical Name or Material (R)-2-Butylsuccinic acid-1-methyl ester
Name Note 95%, (98% e.e.)
CAS 850411-21-1
CAS Min % 94.0
CAS Max % 100.0
Assay ≥94%
Assay Percent Range 95%
Molecular Formula C9 H16 O4
Formula Weight 188.2
MDL Number MFCD03427672