missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

(R)-2-Butylsuccinic acid-1-methyl ester, 95%, 98% e.e., ACROS Organics™

$1,103.33 - $1,103.33

Chemical Identifiers

CAS 850411-21-1
CAS Min % 94.0
CAS Max % 100.0
Assay Percent Range 95%
Molecular Formula C9 H16 O4
Formula Weight 188.2
MDL Number MFCD03427672
Chemical Name or Material (R)-2-Butylsuccinic acid-1-methyl ester, 95%, (98% e.e.)
Assay ≥94%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
AC363085000 SDS Thermo Fisher Scientific
363085000
Glass Ampule with Screw-cap 500mg N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC363080010 SDS Thermo Fisher Scientific
363080010
Glass bottle 1g Each for $1,103.33
Specifications Description & Specifications

Chemical Identifiers

850411-21-1
100.0
C9 H16 O4
MFCD03427672
≥94%
94.0
95%
188.2
(R)-2-Butylsuccinic acid-1-methyl ester, 95%, (98% e.e.)

Specifications

188.2
500mg
Glass Ampule with Screw-cap
94.0
95%
MFCD03427672
Authentic
≥94%
Liquid
Clear orange
100.0
C9 H16 O4
850411-21-1
97.5% min. (GC)
(R)-2-Butylsuccinic acid-1-methyl ester, 95%, (98% e.e.)

Safety and Handling

TSCA : TSCA