Please login to your online account to display your discounted pricing

(R)-(+)-2-Bromopropionic acid, 99%, >85% ee, ACROS Organics™


Quantity 5g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  277920050

 View more versions of this product

Catalog No. AC277920050

  • / Each
 

Description & Specifications

Chemical Identifiers

Assay ≥98.5%
Physical Form Liquid
Quantity 5g
Color Clear colorless to light brown
Beilstein 02, 254
Chemical Name or Material (R)-(+)-2-Bromopropionic acid
Health Hazard 2 GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Specific Rotation +24° to +30° (20°C, 589nm) (neat)
CAS 10009-70-8
Refractive Index 1.4740 to 1.4760 (20°C, 589nm)
Infrared Spectrum Authentic
Enantiomeric Excess 85% min.
Formula Weight 152.98
Molecular Formula C3H5BrO2
CAS Max % 100.0
Name Note >85%ee
Assay Percent Range 99%
Health Hazard 3 GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Packaging Glass bottle
CAS Min % 98.5
Health Hazard 1 GHS Signal Word: Danger
Linear Formula CH3CH(Br)CO2H
MDL Number MFCD00145205

Specifications

Chemical Name or Material (R)-(+)-2-Bromopropionic acid
Name Note >85%ee
CAS 10009-70-8
CAS Min % 98.5
CAS Max % 100.0
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C3H5BrO2
Linear Formula CH3CH(Br)CO2H
Formula Weight 152.98
MDL Number MFCD00145205
Accessories

GHS Signal Word: Danger

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.

SDS & Safety Documents