missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
(R)-(+)-2-(Boc-aminomethyl)-4,4-diethoxypiperidine fumarate, Thermo Scientific™
Supplier: Thermo Scientific Chemicals H25909ME
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
NotesStore in a cool, dry place in tightly closed container. Incompatible with oxidizing agents.
Specifications
(R)-(+)-2-(Boc-aminomethyl)-4,4-diethoxypiperidine fumarate | |
C19H34N2O8 | |
500mg | |
HCNKZGJFFYKTGU-SNXORLAUSA-N | |
(E)-but-2-enedioic acid;tert-butyl N-[[(2R)-4,4-diethoxypiperidin-2-yl]methyl]carbamate | |
44119499 |
1301706-76-2 | |
MFCD09953465 | |
r-+-2-boc-aminomethyl-4,4-diethoxypiperidine fumarate, 2r-2-aminomethyl-4,4-diethoxypiperidine fumarate, 2-boc protected, fumaric acid; tert-butyl n-2r-4,4-diethoxypiperidin-2-yl methyl carbamate, tert-butyl 2r-4,4-diethoxypiperidin-2-ylmethyl carbamate 2e-but-2-enedioate | |
CCOC1(CCNC(C1)CNC(=O)OC(C)(C)C)OCC.C(=CC(=O)O)C(=O)O | |
418.487 | |
418.49 |
Chemical Identifiers
1301706-76-2 | |
418.487 | |
HCNKZGJFFYKTGU-SNXORLAUSA-N | |
44119499 | |
CCOC1(CCNC(C1)CNC(=O)OC(C)(C)C)OCC.C(=CC(=O)O)C(=O)O |
C19H34N2O8 | |
MFCD09953465 | |
r-+-2-boc-aminomethyl-4,4-diethoxypiperidine fumarate, 2r-2-aminomethyl-4,4-diethoxypiperidine fumarate, 2-boc protected, fumaric acid; tert-butyl n-2r-4,4-diethoxypiperidin-2-yl methyl carbamate, tert-butyl 2r-4,4-diethoxypiperidin-2-ylmethyl carbamate 2e-but-2-enedioate | |
(E)-but-2-enedioic acid;tert-butyl N-[[(2R)-4,4-diethoxypiperidin-2-yl]methyl]carbamate |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures