missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ (R)-2-Boc-aminobutyric acid dicyclohexylammonium salt, 95%

$106.26 - $422.54

Chemical Identifiers

CAS 27494-47-9
Molecular Formula C21H40N2O4
Molecular Weight (g/mol) 384.561
MDL Number MFCD00235886
InChI Key HOSMYZXDFVUSCV-FCXZQVPUSA-N
Synonym boc-d-abu-oh dcha, boc-d-abu-oh.dcha, dicyclohexylamine r-2-tert-butoxycarbonyl amino butanoate, boc-d-abu-oh dicyclohexylammonium salt, 2r-2-tert-butoxycarbonyl amino butanoic acid; dicha, 2r-2-tert-butoxy carbonyl amino butanoic acid; n-cyclohexylcyclohexanamine, boc-d-abu-ohdcha, boc-d-abu-oh dicyclohexylammonium salt, r-n-t-butyloxycarbonyl-2-aminobutyric acid dicyclohexylamine, boc-d-abu-oh dicyclohexylammonium salt chn
PubChem CID 2755935
IUPAC Name N-cyclohexylcyclohexanamine;(2R)-2-[(2-methylpropan-2-yl)oxycarbonylamino]butanoic acid
SMILES CCC(C(=O)O)NC(=O)OC(C)(C)C.C1CCC(CC1)NC2CCCCC2
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH6623306 View Documents Alfa Aesar™
H6623306
5g Each for $106.26
 
AAH6623314 View Documents Alfa Aesar™
H6623314
25g Each for $422.54
 
Specifications

Chemical Identifiers

27494-47-9
384.561
HOSMYZXDFVUSCV-FCXZQVPUSA-N
2755935
CCC(C(=O)O)NC(=O)OC(C)(C)C.C1CCC(CC1)NC2CCCCC2
C21H40N2O4
MFCD00235886
boc-d-abu-oh dcha, boc-d-abu-oh.dcha, dicyclohexylamine r-2-tert-butoxycarbonyl amino butanoate, boc-d-abu-oh dicyclohexylammonium salt, 2r-2-tert-butoxycarbonyl amino butanoic acid; dicha, 2r-2-tert-butoxy carbonyl amino butanoic acid; n-cyclohexylcyclohexanamine, boc-d-abu-ohdcha, boc-d-abu-oh dicyclohexylammonium salt, r-n-t-butyloxycarbonyl-2-aminobutyric acid dicyclohexylamine, boc-d-abu-oh dicyclohexylammonium salt chn
N-cyclohexylcyclohexanamine;(2R)-2-[(2-methylpropan-2-yl)oxycarbonylamino]butanoic acid

Specifications

White
27494-47-9
MFCD00235886
boc-d-abu-oh dcha, boc-d-abu-oh.dcha, dicyclohexylamine r-2-tert-butoxycarbonyl amino butanoate, boc-d-abu-oh dicyclohexylammonium salt, 2r-2-tert-butoxycarbonyl amino butanoic acid; dicha, 2r-2-tert-butoxy carbonyl amino butanoic acid; n-cyclohexylcyclohexanamine, boc-d-abu-ohdcha, boc-d-abu-oh dicyclohexylammonium salt, r-n-t-butyloxycarbonyl-2-aminobutyric acid dicyclohexylamine, boc-d-abu-oh dicyclohexylammonium salt chn
CCC(C(=O)O)NC(=O)OC(C)(C)C.C1CCC(CC1)NC2CCCCC2
384.561
384.56
95%
5g
C21H40N2O4
6032357
HOSMYZXDFVUSCV-FCXZQVPUSA-N
N-cyclohexylcyclohexanamine;(2R)-2-[(2-methylpropan-2-yl)oxycarbonylamino]butanoic acid
2755935
Powder
(R)-2-Boc-aminobutyric acid dicyclohexylammonium salt
Safety and Handling

Safety and Handling

storageNote1 : 2° to 8°C