missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
(R)-2-(Boc-amino)butyric acid, 95%, Thermo Scientific Chemicals
$70.71 - $983.51
Chemical Identifiers
CAS | 45121-22-0 |
---|---|
Molecular Formula | C9H17NO4 |
Molecular Weight (g/mol) | 203.238 |
MDL Number | MFCD00270335 |
InChI Key | PNFVIPIQXAIUAY-ZCFIWIBFSA-N |
Synonym | boc-d-abu-oh, r-2-tert-butoxycarbonyl amino butanoic acid, r-n-boc-2-aminobutyric acid, boc-d-alpha-aminobutyric acid, boc-r-2-aminobutyric acid, boc-d-2-aminobutyric acid, 2r-2-tert-butoxycarbonyl amino butanoic acid, 2r-2-tert-butoxy carbonyl amino butanoic acid, butanoic acid, 2-1,1-dimethylethoxy carbonyl amino-, 2r, 2-r-tert-butoxycarbonylaminobutyric acid |
PubChem CID | 2755936 |
IUPAC Name | (2R)-2-[(2-methylpropan-2-yl)oxycarbonylamino]butanoic acid |
SMILES | CCC(C(=O)O)NC(=O)OC(C)(C)C |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAH6399103
|
Thermo Scientific Chemicals
H6399103 |
1 g |
Each for $70.71
|
|
|||||
AAH6399106
|
Thermo Scientific Chemicals
H6399106 |
5 g |
Each for $250.60
|
|
|||||
AAH6399114
|
Thermo Scientific Chemicals
H6399114 |
25 g |
Each for $983.51
|
|
|||||
Description
(R)-2-(Boc-amino)butyric acid is used in coupling reaction. Also used in preparation of dyes.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications(R)-2-(Boc-amino)butyric acid is used in coupling reaction. Also used in preparation of dyes.
Solubility
Soluble in DMF (1mmol in 1ml DMF).
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, heat.
Chemical Identifiers
45121-22-0 | |
203.238 | |
PNFVIPIQXAIUAY-ZCFIWIBFSA-N | |
2755936 | |
CCC(C(=O)O)NC(=O)OC(C)(C)C |
C9H17NO4 | |
MFCD00270335 | |
boc-d-abu-oh, r-2-tert-butoxycarbonyl amino butanoic acid, r-n-boc-2-aminobutyric acid, boc-d-alpha-aminobutyric acid, boc-r-2-aminobutyric acid, boc-d-2-aminobutyric acid, 2r-2-tert-butoxycarbonyl amino butanoic acid, 2r-2-tert-butoxy carbonyl amino butanoic acid, butanoic acid, 2-1,1-dimethylethoxy carbonyl amino-, 2r, 2-r-tert-butoxycarbonylaminobutyric acid | |
(2R)-2-[(2-methylpropan-2-yl)oxycarbonylamino]butanoic acid |
Specifications
45121-22-0 | |
C9H17NO4 | |
1 g | |
Soluble in DMF (1mmol in 1ml DMF). | |
CCC(C(=O)O)NC(=O)OC(C)(C)C | |
203.238 | |
203.24 | |
Liquid |
Yellow | |
MFCD00270335 | |
boc-d-abu-oh, r-2-tert-butoxycarbonyl amino butanoic acid, r-n-boc-2-aminobutyric acid, boc-d-alpha-aminobutyric acid, boc-r-2-aminobutyric acid, boc-d-2-aminobutyric acid, 2r-2-tert-butoxycarbonyl amino butanoic acid, 2r-2-tert-butoxy carbonyl amino butanoic acid, butanoic acid, 2-1,1-dimethylethoxy carbonyl amino-, 2r, 2-r-tert-butoxycarbonylaminobutyric acid | |
PNFVIPIQXAIUAY-ZCFIWIBFSA-N | |
(2R)-2-[(2-methylpropan-2-yl)oxycarbonylamino]butanoic acid | |
2755936 | |
95% | |
(R)-2-(Boc-amino)butyric acid |
Safety and Handling
TSCA : No
storageNote1 : -30° to -10°C
Recommended Storage : Store at -20°C
RUO – Research Use Only