missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
(R)-2-(Boc-amino)butyric acid, 95%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals H6399114
Description
(R)-2-(Boc-amino)butyric acid is used in coupling reaction. Also used in preparation of dyes.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications(R)-2-(Boc-amino)butyric acid is used in coupling reaction. Also used in preparation of dyes.
Solubility
Soluble in DMF (1mmol in 1ml DMF).
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, heat.
Specifications
(R)-2-(Boc-amino)butyric acid | |
Yellow | |
MFCD00270335 | |
boc-d-abu-oh, r-2-tert-butoxycarbonyl amino butanoic acid, r-n-boc-2-aminobutyric acid, boc-d-alpha-aminobutyric acid, boc-r-2-aminobutyric acid, boc-d-2-aminobutyric acid, 2r-2-tert-butoxycarbonyl amino butanoic acid, 2r-2-tert-butoxy carbonyl amino butanoic acid, butanoic acid, 2-1,1-dimethylethoxy carbonyl amino-, 2r, 2-r-tert-butoxycarbonylaminobutyric acid | |
PNFVIPIQXAIUAY-ZCFIWIBFSA-N | |
(2R)-2-[(2-methylpropan-2-yl)oxycarbonylamino]butanoic acid | |
2755936 | |
95% |
45121-22-0 | |
C9H17NO4 | |
25 g | |
Soluble in DMF (1mmol in 1ml DMF). | |
CCC(C(=O)O)NC(=O)OC(C)(C)C | |
203.238 | |
203.24 | |
Liquid |
Chemical Identifiers
45121-22-0 | |
203.238 | |
PNFVIPIQXAIUAY-ZCFIWIBFSA-N | |
2755936 | |
CCC(C(=O)O)NC(=O)OC(C)(C)C |
C9H17NO4 | |
MFCD00270335 | |
boc-d-abu-oh, r-2-tert-butoxycarbonyl amino butanoic acid, r-n-boc-2-aminobutyric acid, boc-d-alpha-aminobutyric acid, boc-r-2-aminobutyric acid, boc-d-2-aminobutyric acid, 2r-2-tert-butoxycarbonyl amino butanoic acid, 2r-2-tert-butoxy carbonyl amino butanoic acid, butanoic acid, 2-1,1-dimethylethoxy carbonyl amino-, 2r, 2-r-tert-butoxycarbonylaminobutyric acid | |
(2R)-2-[(2-methylpropan-2-yl)oxycarbonylamino]butanoic acid |
Safety and Handling
TSCA : No
storageNote1 : -30° to -10°C
Recommended Storage : Store at -20°C
RUO – Research Use Only