missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ (R)-2-(Boc-amino)-4-phenylbutyric acid, 98%

Manufacturer:  Alfa Aesar™ H6118303

 View more versions of this product

Catalog No. AAH6118303



Description

Description

Applications
(R)-2-(Boc-amino)-4-phenylbutyric acid is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff fields.

Solubility
Slightly soluble in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
Specifications

Specifications

(R)-2-(Boc-amino)-4-phenylbutyric acid
71°C to 78°C
82732-07-8
MFCD00076905
2r-2-tert-butoxycarbonyl amino-4-phenylbutanoic acid
MCODLPJUFHPVQP-GFCCVEGCSA-N
(2R)-2-[(2-methylpropan-2-yl)oxycarbonylamino]-4-phenylbutanoic acid
2755954
Powder
White
1g
C15H21NO4
3653505
Slightly soluble in water.
CC(C)(C)OC(=O)NC(CCC1=CC=CC=C1)C(=O)O
279.336
279.33
98%

Chemical Identifiers

82732-07-8
279.336
MCODLPJUFHPVQP-GFCCVEGCSA-N
2755954
CC(C)(C)OC(=O)NC(CCC1=CC=CC=C1)C(=O)O
C15H21NO4
MFCD00076905
2r-2-tert-butoxycarbonyl amino-4-phenylbutanoic acid
(2R)-2-[(2-methylpropan-2-yl)oxycarbonylamino]-4-phenylbutanoic acid
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Ambient temperatures

Documents