missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ (R)-2-(Boc-amino)-3-(Fmoc-amino)propionic acid, 95%

$44.72 - $582.92

Chemical Identifiers

CAS 131570-56-4
Molecular Formula C23H26N2O6
Molecular Weight (g/mol) 426.469
MDL Number MFCD00236844
InChI Key MVWPBNQGEGBGRF-LJQANCHMSA-N
Synonym boc-d-dap fmoc-oh, boc-n3-fmoc-d-2,3-diaminopropionic acid, r-3-9h-fluoren-9-yl methoxy carbonyl amino-2-tert-butoxycarbonyl amino propanoic acid, n-boc-n'-fmoc-d-2,3-diaminopropionic acid, boc-d-dpr fmoc-oh, ambotzbaa1031, boc-d-dapa fmoc-oh, pubchem14750, rarechem bk pt 0245, boc-d-dapa fmoc-oh.solvent
PubChem CID 2755948
IUPAC Name (2R)-3-(9H-fluoren-9-ylmethoxycarbonylamino)-2-[(2-methylpropan-2-yl)oxycarbonylamino]propanoic acid
SMILES CC(C)(C)OC(=O)NC(CNC(=O)OCC1C2=CC=CC=C2C3=CC=CC=C13)C(=O)O
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH62403MD View Documents Alfa Aesar™
H62403MD
250mg Each for $44.72
 
AAH6240303 View Documents Alfa Aesar™
H6240303
1g Each for $132.77
 
AAH6240306 View Documents Alfa Aesar™
H6240306
5g Each for $582.92
 
Description

Description

Applications
(R)-2-(Boc-amino)-3-(Fmoc-amino)propionic acid is used as pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Store in a cool, dry place, in a well sealed container. Store away from oxidizing agents.
Specifications

Chemical Identifiers

131570-56-4
426.469
MVWPBNQGEGBGRF-LJQANCHMSA-N
2755948
CC(C)(C)OC(=O)NC(CNC(=O)OCC1C2=CC=CC=C2C3=CC=CC=C13)C(=O)O
C23H26N2O6
MFCD00236844
boc-d-dap fmoc-oh, boc-n3-fmoc-d-2,3-diaminopropionic acid, r-3-9h-fluoren-9-yl methoxy carbonyl amino-2-tert-butoxycarbonyl amino propanoic acid, n-boc-n'-fmoc-d-2,3-diaminopropionic acid, boc-d-dpr fmoc-oh, ambotzbaa1031, boc-d-dapa fmoc-oh, pubchem14750, rarechem bk pt 0245, boc-d-dapa fmoc-oh.solvent
(2R)-3-(9H-fluoren-9-ylmethoxycarbonylamino)-2-[(2-methylpropan-2-yl)oxycarbonylamino]propanoic acid

Specifications

250mg
C23H26N2O6
boc-d-dap fmoc-oh, boc-n3-fmoc-d-2,3-diaminopropionic acid, r-3-9h-fluoren-9-yl methoxy carbonyl amino-2-tert-butoxycarbonyl amino propanoic acid, n-boc-n'-fmoc-d-2,3-diaminopropionic acid, boc-d-dpr fmoc-oh, ambotzbaa1031, boc-d-dapa fmoc-oh, pubchem14750, rarechem bk pt 0245, boc-d-dapa fmoc-oh.solvent
CC(C)(C)OC(=O)NC(CNC(=O)OCC1C2=CC=CC=C2C3=CC=CC=C13)C(=O)O
426.469
426.46
(R)-2-(Boc-amino)-3-(Fmoc-amino)propionic acid
131570-56-4
MFCD00236844
MVWPBNQGEGBGRF-LJQANCHMSA-N
(2R)-3-(9H-fluoren-9-ylmethoxycarbonylamino)-2-[(2-methylpropan-2-yl)oxycarbonylamino]propanoic acid
2755948
95%