missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
(R)-2-(Boc-amino)-3-(Fmoc-amino)propionic acid, 95%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals H6240303
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Applications(R)-2-(Boc-amino)-3-(Fmoc-amino)propionic acid is used as pharmaceutical intermediate.
Solubility
Slightly soluble in water.
Notes
Store in a cool, dry place, in a well sealed container. Store away from oxidizing agents.
Specifications
(R)-2-(Boc-amino)-3-(Fmoc-amino)propionic acid | |
C23H26N2O6 | |
1 g | |
Slightly soluble in water. | |
CC(C)(C)OC(=O)NC(CNC(=O)OCC1C2=CC=CC=C2C3=CC=CC=C13)C(=O)O | |
426.469 | |
426.46 |
131570-56-4 | |
MFCD00236844 | |
boc-d-dap fmoc-oh, boc-n3-fmoc-d-2,3-diaminopropionic acid, r-3-9h-fluoren-9-yl methoxy carbonyl amino-2-tert-butoxycarbonyl amino propanoic acid, n-boc-n'-fmoc-d-2,3-diaminopropionic acid, boc-d-dpr fmoc-oh, ambotzbaa1031, boc-d-dapa fmoc-oh, pubchem14750, rarechem bk pt 0245, boc-d-dapa fmoc-oh.solvent | |
MVWPBNQGEGBGRF-LJQANCHMSA-N | |
(2R)-3-(9H-fluoren-9-ylmethoxycarbonylamino)-2-[(2-methylpropan-2-yl)oxycarbonylamino]propanoic acid | |
2755948 | |
95% |
Chemical Identifiers
131570-56-4 | |
426.469 | |
MVWPBNQGEGBGRF-LJQANCHMSA-N | |
2755948 | |
CC(C)(C)OC(=O)NC(CNC(=O)OCC1C2=CC=CC=C2C3=CC=CC=C13)C(=O)O |
C23H26N2O6 | |
MFCD00236844 | |
boc-d-dap fmoc-oh, boc-n3-fmoc-d-2,3-diaminopropionic acid, r-3-9h-fluoren-9-yl methoxy carbonyl amino-2-tert-butoxycarbonyl amino propanoic acid, n-boc-n'-fmoc-d-2,3-diaminopropionic acid, boc-d-dpr fmoc-oh, ambotzbaa1031, boc-d-dapa fmoc-oh, pubchem14750, rarechem bk pt 0245, boc-d-dapa fmoc-oh.solvent | |
(2R)-3-(9H-fluoren-9-ylmethoxycarbonylamino)-2-[(2-methylpropan-2-yl)oxycarbonylamino]propanoic acid |
Safety and Handling
TSCA : No
Recommended Storage : Ambient temperatures