Please login to your online account to display your discounted pricing

(R)-2-(BOC-amino)-1-propanol, 98%, ACROS Organics™


Quantity 1g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC425990010

 View more versions of this product

Catalog No. AC425990010

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 175.23
Quantity 1g
CAS Max % 100.0
Assay Percent Range 98%
Linear Formula HOCH2CH(CH3)NNCO2C(CH3)3
Chemical Name or Material (R)-2-(BOC-Amino)-1-propanol
Molecular Formula C8H17NO3
CAS 106391-86-0
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
MDL Number MFCD00235912
CAS Min % 97.5
Packaging Glass bottle
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.

Specifications

Chemical Name or Material (R)-2-(BOC-Amino)-1-propanol
CAS 106391-86-0
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Molecular Formula C8H17NO3
Linear Formula HOCH2CH(CH3)NNCO2C(CH3)3
Formula Weight 175.23
MDL Number MFCD00235912
Accessories

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.