missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

(R)-2-(BOC-amino)-1-propanol, 98%, ACROS Organics™

Chemical Identifiers

CAS 106391-86-0
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Molecular Formula C8H17NO3
Linear Formula HOCH2CH(CH3)NNCO2C(CH3)3
Formula Weight 175.23
MDL Number MFCD00235912
Chemical Name or Material (R)-2-(BOC-Amino)-1-propanol
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
AC425990010 Thermo Fisher Scientific
AC425990010
Glass bottle 1g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC425990050 Thermo Fisher Scientific
AC425990050
Glass bottle 5g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications

Description

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Description & Specifications

Chemical Identifiers

106391-86-0
100.0
C8H17NO3
175.23
(R)-2-(BOC-Amino)-1-propanol
97.5
98%
HOCH2CH(CH3)NNCO2C(CH3)3
MFCD00235912

Specifications

106391-86-0
100.0
98%
HOCH2CH(CH3)NNCO2C(CH3)3
MFCD00235912
(R)-2-(BOC-Amino)-1-propanol
97.5
Glass bottle
C8H17NO3
175.23
1g

Safety and Handling

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.