Please login to your online account to display your discounted pricing

Alfa Aesar™ (R)-(+)-2-Benzylsuccinic acid 1-methyl ester, 96%, ee 98+%

$266.30 - $798.89

Chemical Identifiers

Chemical Name or Material (R)-(+)-2-Benzylsuccinic acid 1-methyl ester
CAS 119807-84-0
Molecular Formula C12H14O4
Formula Weight 222.24
MDL Number MFCD00211258
Beilstein 8057113
Synonym (R)-(+)-3-Methoxycarbonyl-4-phenylbutyric acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL18316MD

  SDS
alfa aesar™
L18316MD
250mg Each for $266.30

AAL1831603

  SDS
alfa aesar™
L1831603
1g Each for $798.89
Description & Specifications

Specifications

Formula Weight 222.24
Assay Percent Notes ee ≥98%
CAS 119807-84-0
Quantity 250mg
Chemical Name or Material (R)-(+)-2-Benzylsuccinic acid 1-methyl ester
Assay 96%
Molecular Formula C12H14O4
MDL Number MFCD00211258
Beilstein 8057113
Synonym (R)-(+)-3-Methoxycarbonyl-4-phenylbutyric acid
Optical Rotation +24° (c=1 in Methanol)