Please login to your online account to display your discounted pricing

(R)-2-Benzylsuccinic acid-1-methyl ester, 95%, 98% e.e., ACROS Organics™


Quantity 500mg
Packaging Glass bottle

Manufacturer: thermo fisher scientific  363025000

 View more versions of this product

Catalog No. AC363025000

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥94%
Physical Form Viscous Liquid or Waxy Mass
Formula Weight 222.24
Quantity 500mg
Color Yellow
TSCA TSCA
CAS Min % 94.0
CAS Max % 100.0
Molecular Formula C12H14O4
MDL Number MFCD00211258
CAS 119807-84-0
Synonym (R)-3-Methoxycarbonyl-4-phenylbutyric acid
Chemical Name or Material (R)-2-Benzylsuccinic acid-1-methyl ester
Infrared Spectrum Authentic
Packaging Glass bottle
Name Note 95%, (98% e.e.)
Assay Percent Range 95%
Enantiomeric Excess 97.5% min. (HPLC)

Specifications

Chemical Name or Material (R)-2-Benzylsuccinic acid-1-methyl ester
Name Note 95%, (98% e.e.)
CAS 119807-84-0
CAS Min % 94.0
CAS Max % 100.0
Assay ≥94%
Assay Percent Range 95%
Molecular Formula C12H14O4
Formula Weight 222.24
MDL Number MFCD00211258
Synonym (R)-3-Methoxycarbonyl-4-phenylbutyric acid