Please login to your online account to display your discounted pricing

(R)-2-Benzylsuccinic acid-1-methyl ester, 95%, 98% e.e., ACROS Organics™

Chemical Identifiers

Chemical Name or Material (R)-2-Benzylsuccinic acid-1-methyl ester
Name Note 95%, (98% e.e.)
CAS 119807-84-0
CAS Min % 94.0
CAS Max % 100.0
Assay ≥94%
Assay Percent Range 95%
Molecular Formula C12H14O4
Formula Weight 222.24
MDL Number MFCD00211258
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC363025000

  SDS
thermo fisher scientific
363025000
Glass bottle 500mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥94%
Physical Form Viscous Liquid or Waxy Mass
Formula Weight 222.24
Quantity 500mg
Color Yellow
TSCA TSCA
CAS Min % 94.0
CAS Max % 100.0
Molecular Formula C12H14O4
MDL Number MFCD00211258
CAS 119807-84-0
Synonym (R)-3-Methoxycarbonyl-4-phenylbutyric acid
Chemical Name or Material (R)-2-Benzylsuccinic acid-1-methyl ester
Infrared Spectrum Authentic
Packaging Glass bottle
Name Note 95%, (98% e.e.)
Assay Percent Range 95%
Enantiomeric Excess 97.5% min. (HPLC)