Learn More
(R)-(+)-2-Benzyloxypropionic acid, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals L1720514
Description
(R)-(+)-2-Benzyloxypropionic acid is used as a pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications(R)-(+)-2-Benzyloxypropionic acid is used as a pharmaceutical intermediate.
Solubility
Insoluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agents.
Specifications
(R)-(+)-2-Benzyloxypropionic acid | |
51°C to 53°C | |
MFCD06799065 | |
4675524 | |
Insoluble in water. | |
+90° (c=1 in IMS) | |
(2R)-2-phenylmethoxypropanoic acid | |
7000104 | |
98% |
100836-85-9 | |
C10H12O3 | |
25 g | |
r-2-benzyloxy propanoic acid, r-+-2-benzyloxy propionic acid, r-+-2-benzyloxypropionic acid, o-benzyl-d-lactic acid, 2r-2-benzyloxy propanoic acid, propanoic acid, 2-phenylmethoxy-, 2r, r-benzyloxypropionic acid, r-2-benzyloxy-propionic acid, 2r-2-benzyloxy propionic acid, 2r-2-phenylmethoxypropanoic acid | |
XWAVPOFYNPXXEL-MRVPVSSYSA-N | |
CC(C(=O)O)OCC1=CC=CC=C1 | |
180.203 | |
180.2 |
Chemical Identifiers
100836-85-9 | |
180.203 | |
XWAVPOFYNPXXEL-MRVPVSSYSA-N | |
7000104 | |
CC(C(=O)O)OCC1=CC=CC=C1 |
C10H12O3 | |
MFCD06799065 | |
r-2-benzyloxy propanoic acid, r-+-2-benzyloxy propionic acid, r-+-2-benzyloxypropionic acid, o-benzyl-d-lactic acid, 2r-2-benzyloxy propanoic acid, propanoic acid, 2-phenylmethoxy-, 2r, r-benzyloxypropionic acid, r-2-benzyloxy-propionic acid, 2r-2-benzyloxy propionic acid, 2r-2-phenylmethoxypropanoic acid | |
(2R)-2-phenylmethoxypropanoic acid |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only