Please login to your online account to display your discounted pricing

Alfa Aesar™ (R)-(+)-2-Benzyloxypropionic acid, 98%


Quantity 5g

Manufacturer: alfa aesar™  L1720506

 View more versions of this product

Catalog No. AAL1720506

  • / Each
 

Description & Specifications

Chemical Identifiers

Formula Weight 180.2
Chemical Name or Material (R)-(+)-2-Benzyloxypropionic acid
Assay 98%
Molecular Formula C10H12O3
MDL Number MFCD06799065
CAS 100836-85-9
Health Hazard 1 Warning
Quantity 5g
Optical Rotation +90° (c=1 in IMS)
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Beilstein 4675524
Synonym O-Benzyl-D-lactic acid
Melting Point 51° to 53°C

Specifications

Chemical Name or Material (R)-(+)-2-Benzyloxypropionic acid
CAS 100836-85-9
Molecular Formula C10H12O3
Formula Weight 180.2
MDL Number MFCD06799065
Beilstein 4675524
Synonym O-Benzyl-D-lactic acid

Applications
(R)-(+)-2-Benzyloxypropionic acid is used as a pharmaceutical intermediate.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agents.

Accessories

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.