Please login to your online account to display your discounted pricing

Alfa Aesar™ (R)-(-)-2-(Anilinomethyl)pyrrolidine, 98%

Chemical Identifiers

Chemical Name or Material (R)-(-)-2-(Anilinomethyl)pyrrolidine
CAS 68295-45-4
Molecular Formula C11H16N2
Formula Weight 176.26
MDL Number MFCD01862117
UN Number UN2735
Beilstein 4839749
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1904203

 
alfa aesar™
L1904203
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 176.26
Chemical Name or Material (R)-(-)-2-(Anilinomethyl)pyrrolidine
Assay 98%
Molecular Formula C11H16N2
MDL Number MFCD01862117
Beilstein 4839749
Health Hazard 1 Danger
Quantity 1g
Health Hazard 2 GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Health Hazard 3 GHS P Statement
P280-P305+P351+P338-P309-P310
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
CAS 68295-45-4
UN Number UN2735
Boiling Point 120° to 122°C (2mmHg)
Flash Point >110°C (230°F)
Optical Rotation −20° (c=1 in Ethanol)
DOT Information Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, N.O.S.

Applications
It is used in the determination of the absolute configuration of amines and α-amino acids by 1h nmr of (r)-o-aryllactic acid amides.

Solubility
Not miscible or difficult to mix in water.

Notes
Air sensitive. Store away from air, carbon dioxide and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under inert gas.

Safety and Handling

Danger

GHS P Statement
P280-P305+P351+P338-P309-P310
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.