Please login to your online account to display your discounted pricing

(R)-2-Aminooctane, 95%, 98% e.e., ACROS Organics™


Quantity 500mg
Packaging Glass Ampule with Screw-cap

Manufacturer: thermo fisher scientific  363115000

 View more versions of this product

Catalog No. AC363115000

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥94%
Physical Form Liquid
Formula Weight 129.24
Quantity 500mg
Color Clear yellow
Packaging Glass Ampule with Screw-cap
TSCA TSCA
CAS Min % 94.0
CAS Max % 100.0
Chemical Name or Material (R)-2-Aminooctane
Name Note 95%, (98% e.e.)
MDL Number MFCD01091017
Health Hazard 2 GHS H Statement:
Toxic if swallowed.
Causes serious eye damage.
Causes severe skin burns and eye damage.
Flammable liquid and vapor.
Highly flammable liquid and vapor.
Health Hazard 3 GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Assay Percent Range 95%
Molecular Formula C8 H19 N
Linear Formula CH3(CH2)5CH(CH3)NH2
CAS 34566-05-7
Enantiomeric Excess 97.5% min. (GC)
Synonym (2R)-Octylamine
Infrared Spectrum Authentic
Health Hazard 1 GHS Signal Word: Danger

Specifications

Chemical Name or Material (R)-2-Aminooctane
Name Note 95%, (98% e.e.)
CAS 34566-05-7
CAS Min % 94.0
CAS Max % 100.0
Assay ≥94%
Assay Percent Range 95%
Molecular Formula C8 H19 N
Linear Formula CH3(CH2)5CH(CH3)NH2
Formula Weight 129.24
MDL Number MFCD01091017
Synonym (2R)-Octylamine
Accessories

GHS Signal Word: Danger

GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Toxic if swallowed.
Causes serious eye damage.
Causes severe skin burns and eye damage.
Flammable liquid and vapor.
Highly flammable liquid and vapor.

SDS & Safety Documents