missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

(R)-2-Aminooctane, 95%, 98% e.e., ACROS Organics™

Chemical Identifiers

CAS 34566-05-7
CAS Min % 94.0
CAS Max % 100.0
Assay Percent Range 95%
Molecular Formula C8 H19 N
Linear Formula CH3(CH2)5CH(CH3)NH2
Formula Weight 129.24
MDL Number MFCD01091017
Synonym (2R)-Octylamine
Chemical Name or Material (R)-2-Aminooctane, 95%, (98% e.e.)
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
AC363110010 SDS Thermo Fisher Scientific
363110010
Glass bottle 1g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC363115000 SDS Thermo Fisher Scientific
363115000
Glass Ampule with Screw-cap 500mg N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

34566-05-7
100.0
C8 H19 N
129.24
(2R)-Octylamine
≥94%
94.0
95%
CH3(CH2)5CH(CH3)NH2
MFCD01091017
(R)-2-Aminooctane, 95%, (98% e.e.)

Specifications

129.24
1g
100.0
CH3(CH2)5CH(CH3)NH2
34566-05-7
MFCD01091017
Glass bottle
Authentic
≥94%
Liquid
Clear yellow
94.0
C8 H19 N
95%
97.5% min. (GC)
(2R)-Octylamine
(R)-2-Aminooctane, 95%, (98% e.e.)

Safety and Handling

GHS H Statement:
Toxic if swallowed.
Causes serious eye damage.
Causes severe skin burns and eye damage.
Flammable liquid and vapor.
Highly flammable liquid and vapor.

GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

TSCA : TSCA