Please login to your online account to display your discounted pricing

Alfa Aesar™ (R)-(-)-2-Aminononane, ChiPros 99+%, ee 98+%

Chemical Identifiers

Chemical Name or Material (R)-(-)-2-Aminononane
CAS 74069-74-2
Molecular Formula C9H21N
Formula Weight 143.27
MDL Number MFCD03844739
UN Number UN2735
Synonym (R)-1-Methyloctylamine; (2R)-Nonylamine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1931003

 
alfa aesar™
L1931003
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL1931006

 
alfa aesar™
L1931006
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL1931014

 
alfa aesar™
L1931014
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 143.27
Chemical Name or Material (R)-(-)-2-Aminononane
Assay ≥99%
Molecular Formula C9H21N
MDL Number MFCD03844739
CAS 74069-74-2
Synonym (R)-1-Methyloctylamine; (2R)-Nonylamine
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H314-H318-H227
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.
Health Hazard 3 GHS P Statement
P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Assay Percent Notes ee ≥98%
UN Number UN2735
Boiling Point 73°C (19mmHg)
Quantity 1g
Flash Point 71°C (159°F)
Optical Rotation −6° (Neat)
DOT Information Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, N.O.S.
Safety and Handling

Danger

GHS P Statement
P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H314-H318-H227
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.