Please login to your online account to display your discounted pricing

Alfa Aesar™ (R)-(+)-2-(Aminomethyl)pyrrolidine dihydrochloride, 95%

Chemical Identifiers

Chemical Name or Material (R)-(+)-2-(Aminomethyl)pyrrolidine dihydrochloride
CAS 119020-04-1
Formula Weight 173.08
MDL Number MFCD11974899
Synonym (R)-(+)-(2-Pyrrolidinyl)methylamine dihydrochloride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6619803

 
alfa aesar™
H6619803
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6619806

 
alfa aesar™
H6619806
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH66198MD

 
alfa aesar™
H66198MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 173.08
Melting Point 124° to 130°C
Chemical Name or Material (R)-(+)-2-(Aminomethyl)pyrrolidine dihydrochloride
MDL Number MFCD11974899
CAS 119020-04-1
Physical Form Solid
Assay 95%
Sensitivity Moisture Sensitive
Synonym (R)-(+)-(2-Pyrrolidinyl)methylamine dihydrochloride
Color White to yellow to brown
Quantity 1g
Storage Note 1 -30° to -10°C
Safety and Handling

-30° to -10°C