Learn More
(R)-2-(Aminomethyl)-1-ethylpyrrolidine, 97%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 357000010
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
(R)-2-(Aminomethyl)-1-ethylpyrrolidine | |
96.0 | |
0.9290g/mL | |
Authentic | |
Glass bottle | |
1.4655 to 1.4675 | |
1g | |
+ 99.50 | |
UNRBEYYLYRXYCG-SSDOTTSWSA-N | |
[(2R)-1-ethylpyrrolidin-2-yl]methanamine | |
854138 | |
97% |
22795-97-7 | |
100.0 | |
57°C | |
96% min. (GC) | |
C7H16N2 | |
MFCD00671482 | |
0.929 | |
r-1-ethylpyrrolidin-2-yl methanamine, r-+-2-aminomethyl-1-ethylpyrrolidine, r-2-aminomethyl-1-ethylpyrrolidine, 2r-1-ethylpyrrolidin-2-yl methanamine, 2-pyrrolidinemethanamine, 1-ethyl-, 2r, 2r-1-ethyl-2-pyrrolidinemethanamine, 1-ethyl-2-pyrrolidinyl methanamine #, pubchem11067, r-+-2-aminomethyl-n-ethylpyrrolidine | |
CCN1CCCC1CN | |
128.22 | |
128.22 |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage.
May cause respiratory irritation.
Flammable liquid and vapor.
Harmful if swallowed.
Harmful in contact with skin.
GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Wear protective gloves/protective clothing/eye p
GHS Signal Word: Danger
TSCA : TSCA
RUO â Research Use Only